Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MGAT3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA512156
Description
MGAT3 Polyclonal Antibody for Western Blot, IHC (P)
There are believed to be over 100 different glycosyltransferases involved in the synthesis of protein-bound and lipid-bound oligosaccharides. MGAT3 (N-acetylglucosaminyltransferase III) transfers a GlcNAc residue to the beta-linked mannose of the trimannosyl core of N-linked oligosaccharides and produces a bisecting GlcNAc. Expression of this gene may be controlled by a multiple-promoter system.
Specifications
MGAT3 | |
Polyclonal | |
Unconjugated | |
MGAT3 | |
1110038J12Rik; AI848272; Beta-1,4-mannosyl-glycoprotein 4-beta-N-acetylglucosaminyltransferase; GGNT3; GlcNAc-T III; GNT3; GNT-III; mannoside acetyl glucosaminyltransferase 3; mannoside acetylglucosaminyltransferase 3; mannosyl (beta-1,4-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase; MGAT3; N-acetylglucosaminyltransferase III; N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase III; RP5-1104E15.2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17309, 4248 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q09327, Q10470 | |
MGAT3 | |
KLH conjugated synthetic peptide between 72-102 amino acids from the N-terminal region of human MGAT3. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction