Learn More
MGAT4A Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 251091AP
Catalog No. 50-173-4727
Add to cart
Description
Description
This gene encodes a key glycosyltransferase that regulates the formation of tri- and multiantennary branching structures in the Golgi apparatus. The encoded protein, in addition to the related isoenzyme B, catalyzes the transfer of N-acetylglucosamine (GlcNAc) from UDP-GlcNAc in a beta-1,4 linkage to the Man-alpha-1,3-Man-beta-1,4-GlcNAc arm of R-Man-alpha-1,6(GlcNAc-beta-1,2-Man-alpha-1,3)Man-beta-1,4-GlcNAc-beta-1,4-GlcNAc-beta-1-Asn. The encoded protein may play a role in regulating the availability of serum glycoproteins, oncogenesis, and differentiation.
Specifications
Specifications
MGAT4A
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9UM21
GlcNAc T IVa, GNT IV, GNT IVA, MGAT4A
Rabbit
IgG
150 μL
-20°C
Primary
11320
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
MGAT4A
Liquid
MGAT4A
MGAT4A Fusion Protein Ag18956
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human