Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MGC13138 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179709
Description
MGC13138 Polyclonal specifically detects MGC13138 in Human samples. It is validated for Western Blot.Specifications
MGC13138 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
NP_219363 | |
ZNF764 | |
Synthetic peptide directed towards the N terminal of human ZNF764. Peptide sequence: APPLAPLPPRDPNGAGPEWREPGAVSFADVAVYFCREEWGCLRPAQRALY | |
Affinity Purified | |
RUO | |
92595 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
MGC13138, zinc finger protein 764 | |
Rabbit | |
45 kDa | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Rabbit: 79%. | |
Human, Rabbit | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title