Learn More
Abnova™ MGC16703 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Manufacturer:  Abnova™ H00113691T01
Catalog No. 89-952-324
Add to cart
Description
Description
Specifications
Specifications
293T
NM_145042
pCMV-MGC16703 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
MGC16703 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 16.7kDa
Transient overexpression cell lysate was tested with anti-MGC16703 antibody (Mfr. No. H00113691-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human