Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MGC20410 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP17926820UL
Description
MGC20410 Polyclonal specifically detects MGC20410 in Human samples. It is validated for Western Blot.Specifications
MGC20410 | |
Polyclonal | |
Western Blot 1:1000 | |
NP_612465 | |
BATF2 | |
Synthetic peptide directed towards the N terminal of human BATF2. Peptide sequence MHLCGGNGLLTQTDPKEQQRQLKKQKNRAAAQRSRQKHTDKADALHQQHE. | |
20 μL | |
Primary | |
Human | |
IgG |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
basic leucine zipper transcription factor, ATF-like 2, basic leucine zipper transcriptional factor ATF-like 2, B-ATF-2, MGC20410, SARI, suppressor of AP-1 regulated by IFN | |
Rabbit | |
Affinity Purified | |
RUO | |
116071 | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title