Learn More
Aviva Systems Biology Corp TEX47 Antibody - N-terminal region (ARP55494_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP55494_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MGC26647. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MGC26647. Alternate names: ApplicationWB. Species reactivity: Cow, Human
Catalog No. 50-014-61526