Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MGC4172 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$149.00 - $329.00

Specifications

Antigen MGC4172
Immunogen Synthetic peptides corresponding to MGC4172(short-chain dehydrogenase/reductase) The peptide sequence was selected from the middle region of MGC4172. Peptide sequence DGHIININSMSGHRVLPLSVTHFYSATKYAVTALTEGLRQELREAQTHIR.
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP15999320 View Documents Novus BiologicalsSupplier Diversity Partner
NBP15999320UL
20ul Each for $149.00
Add to cart
 
NBP159993 View Documents Novus BiologicalsSupplier Diversity Partner
NBP159993
100 ul Each for $329.00
Add to cart
 
Description

Description

MGC4172 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

MGC4172
Rabbit
Polyclonal
Unconjugated
79154
Affinity Purified
Western Blot
RUO
Synthetic peptides corresponding to MGC4172(short-chain dehydrogenase/reductase) The peptide sequence was selected from the middle region of MGC4172. Peptide sequence DGHIININSMSGHRVLPLSVTHFYSATKYAVTALTEGLRQELREAQTHIR.
Primary
IgG
ARPG836, dehydrogenase/reductase (SDR family) member 11, dehydrogenase/reductase SDR family member 11, FLJ39232, MGC4172, SDR24C1, short chain dehydrogenase/reductase family 24C, member 1, short-chain dehydrogenase/reductase
PBS and 2% Sucrose with 0.09% Sodium Azide
Immunogen affinity purified
Store at -20C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit