Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MGC50722 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP19147220UL

 View more versions of this product

Catalog No. NBP19147220


Add to cart

Description

Description

MGC50722 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

MGC50722
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
MGC50722
Synthetic peptide directed towards the N terminal of human MGC50722. Peptide sequence DPPWAAPHVVGSDDLKEPGPWGKACSLPMWSTGPEARDGDSSVSSGRLSC.
Immunogen affinity purified
RUO
Primary
399693
Western Blot
Western Blot 1:1000
NP_976223
hypothetical MGC50722, hypothetical protein LOC399693
Rabbit
IgG
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit