Learn More
Aviva Systems Biology Corp MGC70924 Antibody - middle region (ARP55934_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP55934_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MGC70924. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MGC70924. Alternate names: ApplicationWB. Species reactivity: Horse, Human, Rat
Catalog No. 50-014-61668