Learn More
Selleck Chemical LLCSupplier Diversity Partner MGCD-265 analog 10mg 875337-44-3  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S136110MG
MGCD-265 is a potent, multi-target and ATP-competitive inhibitor of c-Met and VEGFR1/2/3 with IC50 of 1 nM, 3 nM/3 nM/4 nM, respectively; also inhibits Ron and Tie2. Phase 1/2.
Catalog No. 50-675-9