Learn More
MgcRacGAP Chicken anti-Human, Polyclonal, R&D Systems™
Chicken Polyclonal Antibody has been used in 1 publication
Manufacturer:  R&D Systems AF1424SP
 View more versions of this product
Catalog No. AF1424SP
Add to cart
This item is not returnable. View return policy
Description
Description
MgcRacGAP/RACGAP1 Polyclonal specifically detects MgcRacGAP/RACGAP1 in Human samples. It is validated for Western Blot.
Specifications
Specifications
MgcRacGAP/RACGAP1
LYOPH
Western Blot 0.1 ug/mL
Q9H0H5
GTPase activating protein, HsCYK-4, ID-GAP, KIAA1478, Male germ cell RacGap, MGCRACGAP, MgcRacGAPrac GTPase-activating protein 1, Rac GTPase activating protein 1, RACGAP1
Chicken
IgY
25 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human MgcRacGAP in direct ELISAs and Western blots.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
RACGAP1
E. coli-derived recombinant human MgcRacGAP Met1-Lys632 (Lys518Ser) Accession # Q9H0H5
Immunogen affinity purified
RUO
Primary
29127
Reconstitute at 0.2 mg/mL in sterile PBS.