Learn More
MGLL Mouse anti-Human, Clone: OTI2G8, liquid, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner TA502890
Catalog No. 50-167-3893
Add to cart
Description
Description
Monoglyceride lipase functions together with hormone-sensitive lipase to hydrolyze intracellular triglyceride stores in adipocytes and other cells to fatty acids and glycerol. MGLL may also complement lipoprotein lipase in completing hydrolysis of monoglycerides resulting from degradation of lipoprotein triglycerides.
Specifications
Specifications
MGLL
Monoclonal
Unconjugated
MGLL
Liquid
MGLL
Full length human recombit protein of human MGLL produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI2G8
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q99685
HU-K5, HUK5, MAGL, MGL
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
11343