Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mGluR2 Mouse anti-Human, Clone: 6E4F6, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP261834
Description
mGluR2 Monoclonal antibody specifically detects mGluR2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
mGluR2 | |
6E4F6 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
GLUR2, glutamate metabotropic receptor 2, glutamate receptor, metabotropic 2, GPRC1Bmetabotropic glutamate receptor 2, mGlu2, mGluR2, MGLUR2glutamate receptor homolog | |
Mouse | |
IgG1 | |
Protein G purified | |
Apoptosis, Cancer, GPCR, Metabotropic Glutamate Receptors, Neuronal Cell Markers, Neuroscience, Neurotransmission, Tumor Suppressors, Vision | |
Primary | |
2912 |
ELISA, Flow Cytometry, Western Blot | |
1 mg/mL | |
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
Purified | |
GRM2 | |
Purified recombinant fragment of human GRM2 (AA: extra 414-558) expressed in E. Coli. | |
95.6 kDa | |
0.1 ml | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Monoclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title