Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mGluR3 Mouse anti-Human, Alexa Fluor 700, Clone: 440511, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB46682N100UG
This item is not returnable.
View return policy
Description
mGluR3 Monoclonal antibody specifically detects mGluR3 in Human samples. It is validated for Flow CytometrySpecifications
mGluR3 | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
GLUR3, glutamate receptor, metabotropic 3, GPRC1C, GPRC1Cglutamate metabotropic receptor 3, GRM3, mGlu3, mGluR3, MGLUR3metabotropic glutamate receptor 3 | |
Hek293 cells transfected with human mGLUR3, Accession # Q14832 | |
100 μg | |
Primary | |
Reconstitution default not known | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2a |
Flow Cytometry | |
440511 | |
Flow Cytometry 0.25-1 ug/106 cells | |
Q14832 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2913 | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title