Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mGluR4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  LIFE TECHNOLOGIES 720280

Encompass

Catalog No. 72-028-0


Add to cart

Description

Description

These Polyclonal antibodies are of rabbit origin developed by immunizing animals with proteins or peptides. The polyclonal antibody is purified by affinity purification from the rabbit sera generated after immunizing the rabbits with a specific type of protein or peptide. The purified antibody is tested for its functionality in various relevant research applications. The antibody is developed for Research Use Only and is non-hazardous or non-infectious in nature. This antibody is predicted to react with Monkey, Pig, Rabbit and Cat.

mGluR4 Polyclonal antibody specifically detects mGluR4 in Human samples. It is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Specifications

Specifications

mGluR4
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
0.5 mg/mL
PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.2
Q14833
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
2914
Liquid
Polyclonal
Unconjugated
GRM4
G protein-coupled receptor GRM4; G protein-coupled receptor, family C, group 1, member D; glutamate metabotropic receptor 4; glutamate receptor; glutamate receptor, metabotropic 4; Gprc1d; Grm4; Metabotropic glutamate receptor 4; mGlu4; mGluR4
Peptide corresponding to Human GRM4 (aa 891-912)
Affinity chromatography
RUO
Antibody
Polyclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit