Learn More
mGluR5/1a Mouse anti-Human, Mouse, Rat, Biotin, Clone: S75-33, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259334B
Catalog No. NBP259334B
Add to cart
Description
Description
mGluR5/1a Monoclonal antibody specifically detects mGluR5/1a in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
mGluR5/1a
S75-33
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
Fusion protein amino acids 824-1203 (cytoplasmic C-terminus) of rat mGluR5b
Protein G purified
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Biotin
PBS with 0.05% Sodium Azide
Mouse
IgG1
100 ul
Primary
Detects 130kDa (both mGluR1 and mGluR5).