Learn More
Aviva Systems Biology Corp MGMT Antibody - middle region (ARP42038_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP42038_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MGMT. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MGMT. Alternate names: -ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-62937