Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MGMT Antibody (MT 3.1), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 6 publications
Supplier: Novus Biologicals NB1006920.1ML
Description
MGMT Monoclonal specifically detects MGMT in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MGMT | |
Monoclonal | |
Unconjugated | |
Tissue culture supernatant, containing less than 0.1% sodium azide with Sodium Azide | |
6-O-methylguanine-DNA methyltransferase, EC 2.1.1.63, methylated-DNA--protein-cysteine methyltransferase, methylguanine-DNA methyltransferase, O-6-methylguanine-DNA methyltransferase, O6-methylguanine-DNA methyltransferase, O-6-methylguanine-DNA-alkyltransferase | |
Mouse | |
Tissue culture supernatant | |
RUO | |
Primary | |
This stains all mantle zone lymphocytes and 50% of germinal center lymphocytes. Basaloid epithelial cells of tonsil squamous mucosa were also stained positive with this. | |
Store at 4°C. Do not freeze. | |
IgG1 |
Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot | |
MT 3.1 | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:20 | |
P16455 | |
MGMT | |
Recombinant human MGMT protein. | |
0.1 mL | |
Apoptosis, Cancer, Chromatin Research, Direct Reversal of DNA Damage, DNA Repair, Tumor Suppressors | |
4255 | |
Human | |
Supernatant |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction