Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MGMT Mouse anti-Human, Mouse (Negative), Allophycocyanin/Cy7, Clone: MT 23.2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NB100168APCCY7

Catalog No. N10168APCY7

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

MGMT Monoclonal antibody specifically detects MGMT in Human, Mouse (Negative) samples. It is validated for Flow Cytometry.
Specifications

Specifications

MGMT
MT 23.2
Flow Cytometry
Conjugated, Purified
MGMT
Recombinant human MGMT (O6-methylguanine-DNA methyltransferase) purified from E. coli [UniProt No. P16455]
Protein G purified
Apoptosis, Cancer, Chromatin Research, Direct Reversal of DNA Damage, DNA Repair, Tumor Suppressors
Primary
4255.0
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
6-O-methylguanine-DNA methyltransferase, EC 2.1.1.63, methylated-DNA--protein-cysteine methyltransferase, methylguanine-DNA methyltransferase, O-6-methylguanine-DNA methyltransferase, O6-methylguanine-DNA methyltransferase, O-6-methylguanine-DNA-alkyltransferase
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human, Mouse (Negative)
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit