Learn More
MGMT (O-methylguanine-DNA methyltransferase) Ab-1 Mouse Monoclonal Antibody, Epredia™
Epredia™ MGMT (O-methylguanine-DNA methyltransferase) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.
Supplier: Epredia MS470P1ABX
Description
Cancer chemotherapeutic alkylating agents (e.g. BCNU,) act by inducing formation of lethal cross-links at the O-alkylguanine position in DNA. MGMT transfers alkyl adducts from the O-position of guanine in DNA (prior to cross-link formation) to a cysteine residue in its own sequence, thereby restoring DNA to its intact state. This transfer inactivates the MGMT enzyme and is irreversible; hence the level of MGMT in a cell is directly proportional to the level of DNA-damage it can tolerate. In normal tissues, MGMT acts as a suppressor of mutation and carcinogenesis. Tumors with high levels of MGMT are likely be drug-resistant.Host Species: Mouse
Clone: MT 3.1
Isotype: IgG1
Species Reactivity: Human. Also reacts weakly with Mouse, Rat and Hamster. Others not known.
Epitope: aa 8-221
Immunogen: Purified recombinant human MGMT protein
Molecular Weight: 25kDa
Positive Control: Raji cells, Colon CA
Cellular Localization: Nuclear and cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Specifications
MGMT (O-methylguanine-DNA methyltransferase) Ab-1 | |
Monoclonal | |
1mg/mL | |
Mouse | |
100 μL | |
Enzyme | |
Hamster, Human, Mouse, Rat | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
MT 3.1 | |
Unconjugated | |
Purified recombinant human MGMT protein | |
RUO | |
Primary | |
Concentrated |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.