Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ MGMT (O-methylguanine-DNA methyltransferase) Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with the Thermo Scientific™ MGMT (O-methylguanine-DNA methyltransferase) Ab-1, Mouse Monoclonal Antibody.

$165.00 - $1029.00

Specifications

Antigen MGMT (O-methylguanine-DNA methyltransferase) Ab-1
Clone MT 3.1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Host Species Murine
View More Specs

Products 9
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS470B
View Documents
Thermo Scientific™
MS470B
Biotinylated Ab with BSA and Azide 1mL
Each for $955.50
Add to cart
 
MS470B0
View Documents
Thermo Scientific™
MS470B0
Biotinylated Ab with BSA and Azide 100μL
Each for $232.00
Add to cart
 
MS470B1
View Documents
Thermo Scientific™
MS470B1
Biotinylated Ab with BSA and Azide 500μL
Each for $596.00
Add to cart
 
MS470P
View Documents
Thermo Scientific™
MS470P
Purified Ab with BSA and Azide 1mL
Each for $1,029.00
Add to cart
 
MS470P0
View Documents
Thermo Scientific™
MS470P0
Purified Ab with BSA and Azide 100μL
Each for $165.00
Add to cart
 
MS470P1
View Documents
Thermo Scientific™
MS470P1
Purified Ab with BSA and Azide 500μL
Each for $564.00
Add to cart
 
MS470P1ABX
View Documents
Thermo Scientific™
MS470P1ABX
Ab without BSA/Azide (Frozen) 100μL
Each for $504.00
Add to cart
 
MS470PABX
View Documents
Thermo Scientific™
MS470PABX
Ab without BSA/Azide (Frozen) 200μL
Each for $832.50
Add to cart
 
MS470R7
View Documents
Thermo Scientific™
MS470R7
Ready-to-use for IHC (UVLP) 7mL
Each for $645.50
Add to cart
 
Description

Description

Ensure accurate, reproducible results in immunohistochemistry experiments with the Thermo Scientific™ MGMT (O-methylguanine-DNA methyltransferase) Ab-1, Mouse Monoclonal Antibody.

Cancer chemotherapeutic alkylating agents (e.g. BCNU,) act by inducing formation of lethal cross-links at the O-alkylguanine position in DNA. MGMT transfers alkyl adducts from the O-position of guanine in DNA (prior to cross-link formation) to a cysteine residue in its own sequence, thereby restoring DNA to its intact state. This transfer inactivates the MGMT enzyme and is irreversible; hence the level of MGMT in a cell is directly proportional to the level of DNA-damage it can tolerate. In normal tissues, MGMT acts as a suppressor of mutation and carcinogenesis. Tumors with high levels of MGMT are likely be drug-resistant.

Host Species: Mouse

Clone: MT 3.1

Isotype: IgG1

Species Reactivity: Human. Also reacts weakly with Mouse, Rat and Hamster. Others not known.

Epitope: aa 8-221

Immunogen: Purified recombinant human MGMT protein

Molecular Weight: 25kDa

Positive Control: Raji cells, Colon CA

Cellular Localization: Nuclear and cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

MGMT (O-methylguanine-DNA methyltransferase) Ab-1
Immunohistochemistry (Paraffin), Western Blot
Murine
Enzyme
Purified recombinant human MGMT protein
MT 3.1
Monoclonal
RUO
Primary
IgG1
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit