Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MGMT Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5120050

This item is not returnable.
View return policy
Description
Positive test controls include: Jurkat, Mouse liver, Mouse spleen. The target is usually found in the following locations: Nucleus.
MGMT Polyclonal antibody specifically detects MGMT in Human, Mouse samples. It is validated for Western BlotSpecifications
MGMT | |
Polyclonal | |
Unconjugated | |
MGMT | |
0-6-methylguanine-DNA methyltransferase; 6-O-methylguanine-DNA methyltransferase; Agat; AGT; AI267024; methylated DNA protein cysteine methyltransferase; methylated-DNA--protein-cysteine methyltransferase; methylguanine-DNA methyltransferase; MGMT; O(6)-alkylguanine-DNA alkyltransferase; O-6-alkylguanine-DNA alkyltransferase; O6-alkylguanine-DNA alkyltransferase; O-6-methylguan; O6-methylguanine-DNA methyltranferase; O-6-methylguanine-DNA methyltransferase; O6-methylguanine-DNA methyltransferase; O-6-methylguanine-DNA-alkyltransferase | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-207 of human MGMT (XP_005252739.1). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
0.76 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P16455, P26187 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
17314, 4255 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title