Learn More
MHC CLAS II R RT-1 Mouse anti-Rat, Clone: OX-6, Abnova™
Mouse monoclonal antibody raised against rat MHC Class II RT-1B.
Manufacturer:  Abnova Corporation MAB12636
Catalog No. 89-934-546
Add to cart
Description
Description
Specifications
Specifications
MHC CLAS II R RT-1
OX-6
Mouse monoclonal antibody raised against rat MHC Class II RT-1B.
In PBS, pH 7.2 (0.09% sodium azide).
Mouse
IgG1
250 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Flow Cytometry (1:50-1:100) Immunohistochemistry (Frozen sections) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot The optimal working dilution should be determined by the end user.
Liquid
Rat thymocyte membrane glycoprotein.
Protein G purification
RUO
Primary
Rat