Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MHC Class I Mouse anti-Mouse, Rat, PE-Cyanine7, Clone: 14-4-4S, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP143736PECY7
Description
MHC Class I Monoclonal antibody specifically detects MHC Class I in Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
MHC Class I | |
14-4-4S | |
Flow Cytometry | |
Conjugated, Purified | |
HLA-E | |
The immunogen for this antibody was PE MHC class II. | |
Protein A or G purified | |
Adaptive Immunity, Asthma, Cell Biology, Core ESC Like Genes, Diabetes Research, DNA Repair, DNA replication Transcription Translation and Splicing, Immunology, Major Histocompatibility Complexes, Stem Cell Markers | |
Primary | |
3133.0 |
Flow Cytometry | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
DKFZp686P19218, EA1.2, EA2.1, HLA class I histocompatibility antigen, alpha chain E, HLA class I histocompatibility antigen, E alpha chain, HLA-6.2, HLAE, lymphocyte antigen, major histocompatibility complex, class I, E, MHC, MHC class I antigen E, MHC HLA-E alpha-1, MHC HLA-E alpha-2.1, QA1 | |
Mouse | |
IgG2a Kappa | |
0.1 mL | |
Store at 4°C in the dark. Do not freeze. | |
Monoclonal | |
Mouse, Rat |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title