Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MHC Class I, Mouse anti-Rat, Clone: OX18, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP280848
Description
MHC Class I Monoclonal antibody specifically detects MHC Class I in Rat Samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry-Frozen, Flow (Intracellular), CyTOF, Blocking Assay.Specifications
MHC Class I | |
OX18 | |
Unconjugated | |
PBS | |
DKFZp686P19218, EA1.2, EA2.1, HLA class I histocompatibility antigen, alpha chain E, HLA class I histocompatibility antigen, E alpha chain, HLA-6.2, HLAE, lymphocyte antigen, major histocompatibility complex, class I, E, MHC, MHC class I antigen E, MHC HLA-E alpha-1, MHC HLA-E alpha-2.1, QA1 | |
Mouse | |
IgG1 | |
0.1 mg | |
Adaptive Immunity, Asthma, Cell Biology, Core ESC Like Genes, Diabetes Research, DNA Repair, DNA replication Transcription Translation and Splicing, Immunology, Stem Cell Markers | |
Primary | |
3133 |
ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation | |
1 mg/mL | |
Flow Cytometry 1:50-1:100, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Flow (Intracellular), CyTOF-ready, Block/Neutralize | |
Purified | |
HLA-E | |
Rat spleen glycoproteins | |
Protein G purified | |
RUO | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Monoclonal | |
Rat |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title