Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MHC Class II Mouse anti-Human, Bovine, Canine, Equine, DyLight™ 488, Clone: CVS20, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP234848G

Catalog No. NBP234848G


Add to cart

Description

Description

MHC Class II Monoclonal antibody specifically detects MHC Class II in Human, Bovine, Canine, Equine samples. It is validated for Flow Cytometry, Immunohistochemistry.
Specifications

Specifications

MHC Class II
CVS20
Flow Cytometry, Immunohistochemistry
Conjugated, Purified
HLA-DMA
3132 cells
Protein G purified
Adaptive Immunity, Asthma, B Cell Development and Differentiation Markers, Cell Biology, Diabetes Research, Immune System Diseases, Immunology, Major Histocompatibility Complexes, Transcription Factors and Regulators
Primary
3108.0
Flow Cytometry, Immunohistochemistry
DyLight™ 488
50 mM sodium borate with 0.05% sodium azide
class II histocompatibility antigen, M alpha chain, D6S222E, DMA, HLA class II histocompatibility antigen, DM alpha chain, major histocompatibility complex, class II, DM alpha, MHC class II antigen DMA, Really interesting new gene 6 protein, RING6HLADM
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Bovine, Canine, Equine, Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit