Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MI 2 dihydrochloride

Menin-MLL interaction inhibitor

Manufacturer:  Tocris Bioscience™ 4707/10

 View more versions of this product

Catalog No. 47-071-0


Add to cart
This item is not returnable. View return policy

Description

Description

Menin-MLL interaction inhibitor; binds menin (IC50 = 446 nM). Selectively inhibits proliferation and induces apoptosis of leukemia cells harboring MLL fusion proteins. Also blocks transformation and induces hematopoietic differentiation of MLL-AF9 transformed bone marrow cells.
Specifications

Specifications

4-[4-(4,5-Dihydro-5,5-dimethyl-2-thiazolyl)-1-piperazinyl]-6-propylthieno[2,3-d]pyrimidine dihydrochloride
448.48
Target: Methyltransferase Inhibitors
C18H25N5S2.2HCl
10mg
>98%
Safety and Handling

Safety and Handling

Recommended Storage : Store at −20°C

SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit