Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MIA Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550054

Catalog No. PA550054


Add to cart

Description

Description

The antibody detects endogenous levels of total MIA protein.

Elicits growth inhibition on melanoma cells in vitro as well as some other neuroectodermal tumors, including gliomas.
Specifications

Specifications

MIA
Polyclonal
Unconjugated
MIA
Liquid
MIA
Synthesized peptide derived from C-terminal of human MIA.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q16674, Q61865
Melanoma-derived growth regulatory protein, Melanoma inhibitory activity,
Rabbit
IgG
100 μL
-20°C
Primary
12587, 8190
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit