Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MICALCL Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548944

Catalog No. PA548944

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May cooperate with MAPK1/ERK2 via an intracellular signal transduction pathway in the morphogenetic development of round spermatids to spermatozoa.
Specifications

Specifications

MICALCL
Polyclonal
PBS with 0.09% sodium azide
Q6ZW33
MICAL C-terminal-like protein, ERK2-binding testicular protein 1, Ebitein-1, MICALCL
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
84953
Western Blot
Unconjugated
MICALCL
Liquid
MICALCL
KLH conjugated synthetic peptide between 135-163 amino acids from the N-terminal region of human MICALCL.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit