Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MICALL1 Rabbit anti-Human, DyLight 650, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP270017C
Description
MICALL1 Polyclonal antibody specifically detects MICALL1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).Specifications
MICALL1 | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
DKFZp686M2226, KIAA1668, MICAL-L1, MICAL-like 1, MICAL-like protein 1 | |
Rabbit | |
IgG | |
0.1 mL | |
Store at 4°C in the dark. | |
Polyclonal | |
Human |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Affinity Purified, Conjugated | |
MICALL1 | |
Synthetic peptide made to an internal portion of the human MICALL1 protein (between amino acids 200-280) [UniProt Q8N3F8] | |
Immunogen affinity purified | |
Signal Transduction | |
Primary | |
85377.0 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title