Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MICALL2 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA524826

Catalog No. PIPA524826


Only null left
Add to Cart

Description

Description

MICALL2 Polyclonal Antibody for Western Blot, IHC (P), Flow

Effector of small Rab GTPases which is involved in junctional complexes assembly through the regulation of cell adhesion molecules transport to the plasma membrane and actin cytoskeleton reorganization. Regulates the endocytic recycling of occludins, claudins and E-cadherin to the plasma membrane and may thereby regulate the establishment of tight junctions and adherens junctions. In parallel, may regulate actin cytoskeleton reorganization directly through interaction with F-actin or indirectly through actinins and filamins. Most probably involved in the processes of epithelial cell differentiation, cell spreading and neurite outgrowth. [UniProt].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MICALL2
Polyclonal
Unconjugated
MICALL2
JRAB; Junctional Rab13-binding protein; MICAL like 2; MICALL2; MICAL-L2; MICAL-like 2; MICAL-like protein 2; milk2; Molecule interacting with CasL-like 2
Rabbit
Antigen Affinity Chromatography, Protein A
RUO
79778
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.25 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q8IY33
MICALL2
KLH conjugated synthetic peptide between 841-870 amino acids from the C-terminal region of human MILK2.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.