Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MICB Mouse anti-Human, PE-Cyanine7, Clone: MM0473-3C37, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP211798PECY7
Description
MICB Monoclonal antibody specifically detects MICB in Human samples. It is validated for Flow Cytometry.Specifications
MICB | |
MM0473-3C37 | |
Flow Cytometry | |
Conjugated, Purified | |
MICB | |
Human MICB | |
Protein G purified | |
Biologically Active Proteins | |
Primary | |
4277.0 |
Flow Cytometry | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
MHC class I chain-related protein B, MHC class I mic-B antigen, MHC class I polypeptide-related sequence B, MIC-B, PERB11.2MHC class I-like molecule PERB11.2-IMX, stress inducible class I homolog | |
Mouse | |
IgG1 | |
0.1 mL | |
Store at 4°C in the dark. Do not freeze. | |
Monoclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title