Learn More
Sklar InstrumentsSupplier Diversity Partner Michel Clips, Sklar, .75mm x 1.75mm, Clip, Pointed, Stainless Steel, Disposable. PK/1000. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 241007
Michel Clips, Sklar, .75mm x 1.75mm, Clip, Pointed, Stainless Steel, Disposable. Sklar's Michel Clips may be applied to wounds and removed when healing is complete, or they may be used more temporarily as part of a procedure to stop blood flow. The clips are available in a variety of sizes for different incision locations. Pack of 1000.
Catalog No. 50-143-7993