Alfa Aesar™ Michler's Ketone, 98%

Manufacturer: Alfa Aesar™ A118890E

Quantity 2500g

 View more versions of this product

Catalog No. AAA118890E

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 90-94-8
    Molecular Formula C17H20N2O
    Formula Weight 268.36
    MDL Number MFCD00008312
    Beilstein 790733
    Merck Index 14,6177
    Synonym 4,4'-Bis(dimethylamino)benzophenone
    Chemical Name or Material Michler's Ketone

    Specifications

    Formula Weight 268.36
    Health Hazard 3 GHS P Statement
    P280-P281-P305+P351+P338-P310-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 90-94-8
    Boiling Point >360°C
    Beilstein 790733
    Melting Point 174° to 176°C
    MDL Number MFCD00008312
    Merck Index 14,6177
    Health Hazard 1 Danger
    Quantity 2500g
    Flash Point 220°C (428°F)
    TSCA TSCA
    Molecular Formula C17H20N2O
    Health Hazard 2 GHS H Statement
    H350-H341-H318
    May cause cancer.
    Suspected of causing genetic defects.
    Causes serious eye damage.
    RTECS Number DJ0250000
    Synonym 4,4'-Bis(dimethylamino)benzophenone
    Chemical Name or Material Michler's Ketone
    Assay 98%

    GHS H Statement
    H350-H341-H318
    May cause cancer.
    Suspected of causing genetic defects.
    Causes serious eye damage.

    GHS P Statement
    P280-P281-P305+P351+P338-P310-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit