Learn More
MICL/CLEC12A Rat anti-Mouse, Alexa Fluor™ 405, Clone: 812303R, R&D Systems™
Rat Monoclonal Antibody
Manufacturer:  R&D Systems FAB2950RV100UG
Catalog No. FB2950RV100
Add to cart
This item is not returnable. View return policy
Description
Description
MICL/CLEC12A Monoclonal antibody specifically detects MICL/CLEC12A in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
MICL/CLEC12A
812303R
Flow Cytometry 0.25-1 ug/10^6 cells
Q504P2
CD371, CLEC12A, CLL1, CLL-1, C-type lectin domain family 12, member A, DCAL-2, MICL
Mouse myeloma cell line NS0-derived recombinant mouse MICL/CLEC12A, Thr67-Arg267, Accession No. Q504P2
Protein A or G purified from cell culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects mouse MICL/CLEC12A in direct ELISAs.
Flow Cytometry
Alexa Fluor™ 405
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide.
Purified, Supernatant, Conjugated
Rat
IgG2a
100 μg
Primary
160364.0
Mouse