Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Miconazole, 10mg/mL in DMSO, sterile-filtered, Alfa Aesar™

Click to view available options
Quantity:
5 x 1mL
22916-47-8
C18H14Cl4N2O
416.123
MFCD00216019
BYBLEWFAAKGYCD-UHFFFAOYSA-N
4189
CHEBI:82892
1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]imidazole
C1=CC(=C(C=C1Cl)Cl)COC(CN2C=CN=C2)C3=C(C=C(C=C3)Cl)Cl
22916-47-8
C18H14Cl4N2O
416.123
MFCD00216019
BYBLEWFAAKGYCD-UHFFFAOYSA-N
4189
CHEBI:82892
1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]imidazole
C1=CC(=C(C=C1Cl)Cl)COC(CN2C=CN=C2)C3=C(C=C(C=C3)Cl)Cl
Physical Form Liquid
Concentration 10mg/mL in DMSO
Quantity 5 x 1mL
Chemical Name or Material Miconazole, sterile-filtered

GHS H Statement
H317-H227-H208
May cause an allergic skin reaction.
Combustible liquid.

GHS P Statement
P210-P261-P280-P363-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : NI4770000

storageNote1 : −30° to −10°C