Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Miconazole nitrate, 10 mg/mL in ethanol, sterile-filtered, Alfa Aesar™

Click to view available options
Quantity:
5 x 1mL
22832-87-7
C18H15Cl4N3O4
479.135
MFCD00058161
MCCACAIVAXEFAL-UHFFFAOYSA-N
68553
1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]imidazole;nitric acid
C1=CC(=C(C=C1Cl)Cl)COC(CN2C=CN=C2)C3=C(C=C(C=C3)Cl)Cl.[N+](=O)(O)[O-]
22832-87-7
C18H15Cl4N3O4
479.135
MFCD00058161
MCCACAIVAXEFAL-UHFFFAOYSA-N
68553
1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]imidazole;nitric acid
C1=CC(=C(C=C1Cl)Cl)COC(CN2C=CN=C2)C3=C(C=C(C=C3)Cl)Cl.[N+](=O)(O)[O-]
UN Number UN1987
Physical Form Liquid
Concentration 10mg/mL in ethanol
Quantity 5 x 1mL
Chemical Name or Material Miconazole nitrate, sterile-filtered

GHS H Statement
H225-H371-H317
Highly flammable liquid and vapor.
May cause damage to organs.
May cause an allergic skin reaction.

GHS P Statement
P210-P260-P261-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.

RTECSNumber : NI4771000

storageNote1 : −30° to −10°C