Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Miconazole nitrate, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
50mg
This item is not returnable. View return policy
22832-87-7
C18H15Cl4N3O4
479.135
MCCACAIVAXEFAL-UHFFFAOYSA-N
miconazole nitrate,albistat,andergin,aflorix,conofite,florid,micatin,gyno-monistat,epi-monistat,gyno-daktar
68553
1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]imidazole;nitric acid
C1=CC(=C(C=C1Cl)Cl)COC(CN2C=CN=C2)C3=C(C=C(C=C3)Cl)Cl.[N+](=O)(O)[O-]
This item is not returnable. View return policy
22832-87-7
C18H15Cl4N3O4
479.135
MCCACAIVAXEFAL-UHFFFAOYSA-N
miconazole nitrate,albistat,andergin,aflorix,conofite,florid,micatin,gyno-monistat,epi-monistat,gyno-daktar
68553
1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]imidazole;nitric acid
C1=CC(=C(C=C1Cl)Cl)COC(CN2C=CN=C2)C3=C(C=C(C=C3)Cl)Cl.[N+](=O)(O)[O-]

Antifungal agent

Antifungal agent. Inhibits lanosterol demethylase and induces formation of reactive oxygen species.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 50mg
Formula Weight 479.14
Percent Purity >99%
Chemical Name or Material Miconazole nitrate

Recommended Storage : Store at Room Temperature