Learn More
Simport™ Scientific Micrewtube™ Plain Microcentrifuge Tubes
Micrewtube™ Plain Microcentrifuge Tubes can withstand temperatures from -196°C to +121°C.
$150.00
Specifications
Certifications/Compliance RNase, DNase, Pyrogen and DNA-free
Autoclavable Yes
Color Natural
Material Polypropylene
No. per Pack 1000 Pk.
View More Specs
Products 1
Catalog Number Mfr. No. Volume (Metric) dimProductType Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) dimProductType Price Quantity & Availability  
23-045-260
View Documents
Simport™ Scientific
T3414T
1.5 mL
Pack of 1000 for $150.00
Add to Cart
Description
Description
  • Made of polypropylene
  • Autoclavable
  • Certified RNase, DNase, DNA and Pyrogen free
  • Max force: 17,000 g
  • Will withstand temperatures from -196°C to +121°C
Specifications
Specifications
RNase, DNase, Pyrogen and DNA-free
Natural
1000 Pk.
No
Yes
Polypropylene
Non-sterile