Learn More
Micro BlueMaxx™ Light Source Products for Science Education
Provides just the right amount of illumination for searching small areas.
Manufacturer:  Sirchie Finger Print BMMB100
Catalog No. S99190
Add to cart
Includes: Three AAAA alkaline batteries and leather carrying case.
Description
Description
  • Powerful LED blue light source housed in a durable metal body
  • Orange barrier filter slips easily onto the body of the light
  • LED activated with a push-button for momentary use or twist the cap for a steady beam
Specifications
Specifications
Three aaaa alkaline batteries and 7 x 3.25 in., leather carrying case, powerful led blue light source housed in a durable metal body
Provides just the right amount of illumination for searching small areas
Micro Bluemaxx™ Light Source