Learn More
Thermo Scientific™ Micro-Cassette
Easily process your smallest specimens with the Thermo Scientific™ Micro-Cassette, constructed entirely of stainless steel.
Manufacturer:  Thermo Scientific™ 33124
 View more versions of this product
Catalog No. 33-124
  Check Availability
Add to cart
Specifications
Description
Easily process your smallest specimens with the Thermo Scientific™ Micro-Cassette, constructed entirely of stainless steel.
Highlights:
  • Positive clip-on cover for micro specimen
  • Available with fine or coarse mesh
Dimensions:
  • 0.79 x 0.197 in. (20 x 5mm)
Description & Specifications
Specifications
Fine Mesh
5mm
0.79 in.
0.197 in.
Stainless Steel
20mm