Learn More
Micro Essential Lab Insta-Chek™ 0-13 Wide-Range pH Test Paper/Dispenser
Direct reading pH paper with 14 colors for 14 pH values—the widest range available on a single paper
Supplier: Micro Essential Lab JR113
Description
Direct reading pH paper with 14 colors for 14 pH values—the widest range available on a single paper
- Immediate response—colors are distinct and easily read for accurate, reproducible readings
- Single roll dispenser with one 50 ft. × 0.5 in. roll
Order Info
Single-roll dispenser with one 50 ft. long (15.2m) roll of 0 to 13pH paper and color chart; refills sold separately.
Specifications
0 to 13 | |
1000 Tests | |
50 ft. |
Colorimetric chart | |
Jumbo Roll | |
4.57 m |