Micro Essential Lab pH Test Paper Refills for Hydrion™ Dispensers

Manufacturer: Micro Essential Lab 434

 View more versions of this product

Catalog No. 14-853-79

  Check Availability
Add to Cart

    Specifications

    For Use With (Equipment) For Hydrion™ dispensers
    Includes Colorimetric chart
    Quantity 5 Pack
    Product Type Single Roll pH Test Paper Refill
    pH Range 4.5 to 7.5

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit