Learn More
Foxx Life Sciences EZMicro™ Microbiology Funnel Monitor, Gamma Sterilized with 0.2µm Gridded White Removable MCE Membrane, 100mL, PK/50 DFS Item
Manufacturer:  Foxx Life Sciences 3501A36FLS
Foxx Life Sciences disposable Gamma Sterilized EZMicro™ Microbiology Funnel is perfect for any microbiological analysis. These gridded, MCE membranes installed into each unit can reduce up to 70% of analysis time. Each unit is designed to reduce the risk of cross-contamination and are Gamma Irradiated to ensure the highest quality test environment.EZMicro™ Microbiology Funnel Monitor units come with cellulose pad, 100 ml funnel and cover with magnifying glass for colony count. Membrane not welded to the base, it allows to remove it for its culture with optional means and to conserve it as evidence, or to use with ampules of Microclar culture media.
Catalog No. 50-172-0656