Learn More
Roboz SurgicalSupplier Diversity Partner MICRO KDISSECTING TWEEZERS DFS Item
Manufacturer:  Roboz SurgicalSupplier Diversity Partner RS4900
Dumont Tweezers; Pattern #1; .10 X .06mm Tip Size; Stainless Steel; 120mm Length; Material: Inox; Biologie Tips
Catalog No. NC0091017