Cadence Science™ Micro-Mate™ Interchangeable Syringes
Syringe barrels and plungers fit perfectly, eliminating the task of matching up syringe parts
Manufacturer: Cadence Science™ 5014
 View more versions of this product
Catalog No. 14-825-2A
    N/A
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 
    Specifications
    Volume (Metric) 5mL
    Connection Type Luer Tip
    For Use With (Equipment) For micro mate interchangeable barrel of that same size
    Graduations 0.2mL
    Material Borosilicate Glass
    Termination Metal male luer-lock
    Capacity (Metric) 5cc
    Type Interchangeable Syringe
  • Description & Specifications
Related Products
Product Content Feedback