Cadence Science™ Micro-Mate™ Interchangeable Syringes
Syringe barrels and plungers fit perfectly, eliminating the task of matching up syringe parts
Manufacturer: Cadence Science™ 5036
 View more versions of this product
Catalog No. 14-825-9C
    N/A
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 
    Specifications
    Volume (Metric) 20mL
    For Use With (Equipment) For micro mate interchangeable barrel of that same size
    Graduations 1mL
    Material Borosilicate Glass
    Termination Glass male luer-slip
    Capacity (Metric) 20cc
    Type Interchangeable Syringe
  • Description & Specifications
Related Products
Product Content Feedback