Learn More
Cadence Science™ Micro-Mate™ Interchangeable Syringes
Syringe barrels and plungers fit perfectly, eliminating the task of matching up syringe parts
Manufacturer:  Cadence Science™ 5059
 View more versions of this product
Catalog No. 14-825-11C
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
For micro mate interchangeable barrel of that same size
Borosilicate Glass
Interchangeable Syringe
2mL
Glass male luer-slip
50mL