Cadence Science™ Micro-Mate™ Interchangeable Syringes
Syringe barrels and plungers fit perfectly, eliminating the task of matching up syringe parts
Manufacturer: Cadence Science™ 5025
 View more versions of this product
Catalog No. 14-825-3B
    N/A
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 
    Specifications
    Type Interchangeable Syringe
    Volume (Metric) 10mL
    For Use With (Equipment) For micro mate interchangeable barrel of that same size
    Graduations 0.2mL
    Material Borosilicate Glass
    Termination Metal male luer-slip
    Interchangeable syringes are manufactured to ensure the perfect mating of barrels and plungers. Any Micro-Mate interchangeable syringe of a particular size will fit any Micro-Mate interchangeable barrel of that same size.
    • Metal Male Luer Lock, Metal Male Luer or Glass Male Luer tips
    • Constructed of borosilicate glass for heat and corrosion resistance
    • Permanently fused graduations Reinforced flanges with two flat sides prevent syringes from rolling
  • Description & Specifications
Related Products
Product Content Feedback