Learn More
Sklar InstrumentsSupplier Diversity Partner Micro Ophthalmic Basket, Sklar, 13 in. x 10 in. x 1-1/2 in. Micro Ophthalmic Basket Locking Lid, 340mm x 250mm x 40mm, with Silicone Pads, Perforated Sides. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 106660
Micro Ophthalmic Basket, Sklar, 13 in. x 10 in. x 1-1/2 in. Micro Ophthalmic Basket Locking Lid, 340mm x 250mm x 40mm, with Silicone Pads, Perforated Sides. Sklar's Micro Ophthalmic Basket is designed for use in washing, handling and sterilizing small instruments. This product is available in multiple sizes and features a locking lid. The sides are perforated and includes silicone pads to position and stabilize instruments.
Catalog No. 50-143-7265